Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Tolidine 0.1% (w/v) 2 N in HCl

Supplier: TCI America
Synonyms: 4,4'-Bi-o-toluidine

T0255-500ML
TCT0255-500MLEA 42.75 USD InStock
TCT0255-500ML
o-Tolidine 0.1% (w/v) 2 N in HCl
o-Tolidine
Color: Colorless
Form: Clear Liquid

Other Product Names / Synonyms: 4,4'-Diamino-3,3'-dimethylbiphenyl Solution


Formula: [-C₆H₃(CH₃)-₄-NH₂]₂
MW: 212.29 g/mol
MDL Number: MFCD00014773
CAS Number: 119-93-7
UN: 3077
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/45/58/28994558.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty