Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine ≥98.0%

Supplier: TCI America
Synonyms: 2,4,6-Tris(perfluoropropyl)-1,3,5-triazine

T0859-0.1ML
TCT0859-0.1MLEA 231.19 USD InStock
TCT0859-0.1ML
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine ≥98.0%
2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
CAS Number: 915-76-4
MDL Number: MFCD00042438
Molecular Formula: C12F21N3
Molecular Weight: 585.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Specific Gravity (20/20): 1.72

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2,4,6-Tris(perfluoropropyl)-1,3,5-triazine


Formula: C₁₂F₂₁N₃
MW: 585.12 g/mol
Boiling Pt: 165 °C
Density: 1.72 (20 °C)
MDL Number: MFCD00042438
CAS Number: 915-76-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/66/28994966.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty