Synonyms:
Cyclooctatetraene Iron Tricarbonyl
, Cyclooctatetraene iron tricarbonyl
T1775-1G
TCT1775-001GEA
219.09
USD
InStock
TCT1775-001G
Tricarbonyl(cyclooctatetraene)iron ≥96.0% (by titrimetric analysis)
Tricarbonyl(cyclooctatetraene)iron
CAS Number: 12093-05-9
MDL Number: MFCD00001755
Molecular Formula: C11H8FeO3
Molecular Weight: 244.03
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Color: Deep Red
Other Product Names / Synonyms: Cyclooctatetraene Iron Tricarbonyl