Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 4-(Trifluoromethoxy)benzene-1-sulfonyl chloride4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

T2146-5G T2146-25G
TCT2146-25GEA 74.08 USD InStock
TCT2146-25G TCT2146-5G
4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-(Trifluoromethoxy)benzenesulphonyl chloride
CAS Number: 94108-56-2
MDL Number: MFCD00042408
Molecular Formula: C7H4ClF3O3S
Molecular Weight: 260.61
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 113
Specific Gravity (20/20): 1.55


Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 122 °C
Density: 1.55 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00042408
CAS Number: 94108-56-2
UN: 3265
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/07/28996007.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty