Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrakis(triphenylphosphine)platinum(0)

Supplier: TCI America

T2485-1G
TCT2485-1GEA 107.59 USD InStock
TCT2485-1G
Tetrakis(triphenylphosphine)platinum(0)
Tetrakis(triphenylphosphine)platinum (0)
CAS Number: 14221-02-4
MDL Number: MFCD00010014
Molecular Formula: C72H60P4Pt
Molecular Weight: 1244.25
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: 0-10°C


Formula: C₇₂H₆₀P₄Pt
MW: 1244.24 g/mol
MDL Number: MFCD00010014
CAS Number: 14221-02-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/18/28996218.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty