Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraphenylsilane ≥97.0%

Supplier: TCI America

T2554-5G T2554-1G
TCT2554-5GEA 452.55 USD InStock
TCT2554-5G TCT2554-1G
Tetraphenylsilane ≥97.0%
Tetraphenylsilane
CAS Number: 1048-08-4
MDL Number: MFCD00014069
Molecular Formula: C24H20Si
Molecular Weight: 336.51
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 166
Melting point (°C): 238
Flash Point (°C): 193


Formula: C₂₄H₂₀Si
MW: 336.51 g/mol
Boiling Pt: 166 °C
Melting Pt: 238 °C
Flash Pt: 193 °C
MDL Number: MFCD00014069
CAS Number: 1048-08-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/72/28996272.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty