Would you like to log in again?
Purge Adapter, Ace Glass Incorporated
Supplier: Ace Glass
3953-05 3953-03
71001-500EA 144.87 USD InStock
71001-500 71001-502
Purge Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This glass purge adapter is desgined for use with rotary evaporators.
  • Top spherical socket joint
  • Bottom spherical ball joint
  • Middle tube stopcock or valve
  • Fabricated from borosilicate glass
Top has a spherical 35/25 socket joint for joining to the condenser or trap, and the bottom has a spherical 35/25 ball joint for joining to the receiver flask. Middle tube has either a 4mm PTFE stopcock, or a 2mm valve to allow for purging of the vapor from the receiver flask or for adding purge gas into the flask. Overall length is 75mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty