Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(n-propyltetramethylcyclopentadienyl)barium

Supplier: Thermo Scientific Chemicals
Danger

38574.04 38574-04
AAA38574-04EA 436.99 USD InStock
CAAAA38574-04 AAA38574-04
Bis(n-propyltetramethylcyclopentadienyl)barium
Bis(n-propyltetramethylcyclopentadienyl)barium
Viscous liquid/ solid, Packaged in ampoules


Formula: C₂₄H₃₈Ba
Storage Temperature: Ambient
MDL Number: MFCD00151385
CAS Number: 210758-43-3
UN: 1564
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty