Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

B-Tribromoborazine ≥96%

Supplier: Thermo Scientific Chemicals
Danger

43723.06 43723.03 43723-06 43723-03
AAA43723-03EA 115.99 USD InStock
CAAAA43723-03 CAAAA43723-06 AAA43723-03 AAA43723-06
B-Tribromoborazine ≥96%
B-Tribromoborazine
Powder


Formula: B₃Br₃H₃N₃
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00058924
CAS Number: 13703-88-3
UN: 1759
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty