Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraammineplatinum(II) hydrogen phosphate solution (0,5% Pt) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

44076-NH 44076-NB 044076.NB 44076-NH
AAA44076-NBEA 1000.99 USD InStock
CAAAA44076-NB AAA44076-NB AAA44076-NH CAAAA44076-NH
Tetraammineplatinum(II) hydrogen phosphate solution (0,5% Pt) (w/w)
Tetraammineplatinum(II) hydrogen phosphate
Liquid


Formula: H₁₃N₄O₄PPt
MW: 359.18 g/mol
Density: 1,013
Storage Temperature: Ambient
MDL Number: MFCD03788258
CAS Number: 127733-98-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty