Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium 10% on activated carbon powder, eggshell, reduced

Supplier: Thermo Scientific Chemicals
Warning

44696-18 44696.18
AAA44696-18EA 1511.99 USD InStock
CAAAA44696-18 AAA44696-18
Palladium 10% on activated carbon powder, eggshell, reduced
Palladium


Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty