Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol methyl ether acrylate, M.W. 1,000

Supplier: Thermo Scientific Chemicals
Synonyms: mPEG-acrylatePolyethylene glycol monomethyl ether acrylate

046537-06 46537-MD 046537-03
AA46537-06EA 379.99 USD InStock
AA46537-06 AA46537-MD AA46537-03
Polyethylene glycol methyl ether acrylate, M.W. 1,000
Polyethylene glycol methyl ether acrylate
Granular solid


Formula: CH₃(OCH₂CH₂)nOCOC=CH₂
Storage Temperature: Refrigerator
MDL Number: MFCD00803685
CAS Number: 32171-39-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty