Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 4-(4-dimethylaminophenylazo)benzoatep-Methyl red

H27830-06
AAH27830-06EA 199.99 USD InStock
AAH27830-06
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt indicator
4-(4-Dimethylaminophenylazo)benzoic acidsodium salt


Formula: C₁₅H₁₄N₃O₂Na
MW: 291.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00020350
CAS Number: 845-46-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty