Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biphenyl diphenyl ether eutectic

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dowtherm® ADiphyl

H30347-0F H30347-AU
AAH30347-0FEA 287.99 USD InStock
AAH30347-0F AAH30347-AU
Biphenyl diphenyl ether eutectic
Biphenyl diphenyl ether eutectic


Formula: C₂₄H₂₀O
MW: 324.423 g/mol
Boiling Pt: 257 °C
Melting Pt: 12…14 °C
Density: 1.056
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00148859
CAS Number: 8004-13-5
EINECS: 616-827-0
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty