Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia 7 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

H30382-AU H30382-AE
AAH30382-AUEA 439.62 USD InStock
AAH30382-AU AAH30382-AE
Ammonia 7 M in methanol
Ammonia


Formula: NH₃
MW: 17.03 g/mol
Density: 0.779
Flash Pt: 14 °C(57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011418
CAS Number: 7664-41-7
EINECS: 231-635-3
UN: 3286
ADR: "3",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty