Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-2-methylbenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

H32364-03 H32364-06
AAH32364-03EA 85.39 USD InStock
AAH32364-03 AAH32364-06
4-Fluoro-2-methylbenzyl alcohol 99%
4-Fluoro-2-methylbenzyl alcohol


Formula: C₈H₉FO
MW: 140.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701058
CAS Number: 80141-91-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty