Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Heptane 95%

Supplier: Thermo Scientific Chemicals
Danger

H33987-AU H33987-0F
AAH33987-0FEA 82.89 USD InStock
AAH33987-0F AAH33987-AU
n-Heptane 95%
n-Heptane


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 97…99 °C
Melting Pt: –91 °C
Density: 0.684
Flash Pt: -4 °C(25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: "3",II
Merck Index: 13,04679
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty