Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyinosine 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

H52292-03 H52292-06 H52292-MD
AAH52292-03EA 50.29 USD InStock
AAH52292-03 AAH52292-06 AAH52292-MD
2'-Deoxyinosine 98+%
2'-Deoxyinosine


Formula: C₁₀H₁₂N₄O₄
MW: 252.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005762
CAS Number: 890-38-0
EINECS: 212-964-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty