Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine3-Amino-2-methoxypyridine-5-boronic acid Pinacol ester2-(5-Amino-6-methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane2-Methoxy-3-aminopyridine-5-boronic acid pinacol ester

H54570-03 H54570-MD
AAH54570-MDEA 124.32 USD InStock
AAH54570-MD AAH54570-03
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester 96%
3-Amino-2-methoxypyridine-5-boronic acid pinacol ester


Formula: C₁₂H₁₉BN₂O₃
MW: 250.10 g/mol
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD12923427
CAS Number: 893440-50-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty