Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Azidomethyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

H58401-03 H58401-06
AAH58401-06EA 307.99 USD InStock
AAH58401-06 AAH58401-03
4-(Azidomethyl)benzeneboronic acid pinacol ester 95%
4-(Azidomethyl)benzeneboronic acid pinacol ester


Formula: C₁₃H₁₉BN₃O₂
MW: 260.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20922785
CAS Number: 1239481-05-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty