Would you like to log in again?
L-Phenylalanyl-L-phenylalanine
Supplier: Thermo Scientific
J60043-06 J60043-06 J60043-03 J60043-03
AAJ60043-03EA 162 USD InStock
AAJ60043-03 CAAAJ60043-06 CAAAJ60043-03 AAJ60043-06
L-Phenylalanyl-L-phenylalanine
Proteins and Peptides
Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation
Powder
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty