Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Melanocyte-Stimulating Hormone-Release Inhibiting Factor

Supplier: Thermo Scientific
J60045-03 J60045-03 J60045-MD J60045-MD
AAJ60045-MDEA 61.58 USD InStock
AAJ60045-MD AAJ60045-03 CAAAJ60045-MD CAAAJ60045-03
Melanocyte-Stimulating Hormone-Release Inhibiting Factor
Proteins and Peptides
Lyophilized powder

CAS No.: 2002-44-0
Molecular Formula: C13H24N4O3
Formula Weight: 284.36
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00037866

Applications: Blocks the release of alpha-melanocyte stimulating hormone, increases brain dopamine levels and antagonizes physiological and behavioral opioid effects in vivo

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty