Would you like to log in again?
Elastase Substrate V
Supplier: Thermo Scientific
J60093-MB J60093-MB
AAJ60093-MBEA 304.32 USD InStock
AAJ60093-MB CAAAJ60093-MB
Elastase Substrate V
Proteins and Peptides
Fluorogenic substrate for quantification of human leukocyte and porcine pancreatic elastase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty