Would you like to log in again?
Glucose-6-Phosphate Dehydrogenase
Supplier: Thermo Scientific
J60117-JPL J60117-4I J60117-JPL J60117-4I
AAJ60117-JPLEA 1081.22 USD InStock
CAAAJ60117-JPL AAJ60117-JPL CAAAJ60117-4I AAJ60117-4I
Glucose-6-Phosphate Dehydrogenase
Enzymes
Oxidizes glucose-6-phosphate in the presence of NADP+ to yield 6-phosphogluconate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty