Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutralization solution

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DNA Neutralizing solutionNeutralization solutionNeutralisation solutionDNA Neutralising solution

J60275-AP J60275-AP J60275-AK J60275-AK
AAJ60275-AKEA 40.53 USD InStock
AAJ60275-AK CAAAJ60275-AP AAJ60275-AP CAAAJ60275-AK
Neutralization solution
Neutralization solution
Applications: Western Blot and ELISA Reagents
This solution contains: 1.5M NaCl and 1M Tris, pH range 7.0-7.2.
Physical Form: Liquid


Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty