Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

C2 Ceramide 98%

Supplier: Thermo Scientific Chemicals

J60534-MA J60534-LB0 J60534-LB0 J60534-MCR J60534-MCR J60534-MA
AAJ60534-MCREA 31.2 USD InStock
AAJ60534-MCR CAAAJ60534-MA CAAAJ60534-MCR AAJ60534-LB0 AAJ60534-MA CAAAJ60534-LB0
C2 Ceramide 98%
C2 Ceramide
Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases


Formula: C₂₀H₃₉NO₃
MW: 341.536 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Freezer
MDL Number: MFCD00153903
CAS Number: 3102-57-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty