Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dantrolene sodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 3-(((5-(4-nitrophenyl)furan-2-yl)methylene)amino)-2,5-dioxoimidazolidin-1-ide

J60887-MC J60887-MD J60887-MC
AAJ60887-MCEA 111.84 USD InStock
AAJ60887-MC 10814-238 CAAAJ60887-MC
Dantrolene sodium salt
Dantrolene sodium salt
Inhibits intracellular calcium release from the sarcoplasmic reticulum

Crystalline powder


Formula: C₁₄H₉N₄NaO₅
MW: 336.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079130
CAS Number: 14663-23-1
EINECS: 238-706-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty