Would you like to log in again?
Clostripain
Supplier: Thermo Scientific
J61362-MCR J61362-MA J61362-MCR J61362-MA
AAJ61362-MCREA 69.5 USD InStock
AAJ61362-MCR AAJ61362-MA CAAAJ61362-MA CAAAJ61362-MCR
Clostripain
Enzymes
A protease that cleaves proteins on the carboxyl bond of arginine
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty