Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Furegrelate sodium salt ≥99%

Supplier: Thermo Scientific Chemicals

J61378-MF J61378-MA J61378-MF J61378-LB0 J61378-LB0 J61378-MA
AAJ61378-MAEA 230.33 USD InStock
CAAAJ61378-MF CAAAJ61378-LB0 CAAAJ61378-MA AAJ61378-MA AAJ61378-MF AAJ61378-LB0
Furegrelate sodium salt ≥99%
Furegrelate sodium salt
Inhibitor of thrombaxane synthetase

Crystalline solid


Formula: C₁₅H₁₀NNaO₃
Storage Temperature: Freezer
MDL Number: MFCD00058448
CAS Number: 85666-17-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty