Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Epigallocatechin

Supplier: Thermo Scientific Chemicals
Synonyms: 3,5,7,3',4',5'-Hexahydroxylflavan(2R,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol

J61630-MA J61630-MA
AAJ61630-MAEA 191.71 USD InStock
CAAAJ61630-MA AAJ61630-MA
(-)-Epigallocatechin
(-)-Epigallocatechin
A natural product from green tea that induces apoptosis

Powder


Formula: C₁₅H₁₄O₇
MW: 306.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00075939
CAS Number: 970-74-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty