Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Epigallocatechin gallate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl GallateEGCG

J61745-MF J61745-MF
AAJ61745-MFEA 166.02 USD InStock
CAAAJ61745-MF AAJ61745-MF
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate
An antioxidant that acts as an anti-promoter of carcinogenesis, inhibits angiogenesis and induces apoptosis

Powder


Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075940
CAS Number: 989-51-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty