Would you like to log in again?
BLOTTO Antifoam in PBS, with 0.02% Sodium Azide, Thermo Scientific
Supplier: Thermo Scientific
J61888-AK J61888-AK J61888-AP J61888-AP
AAJ61888-APEA 86.88 USD InStock
CAAAJ61888-AK AAJ61888-AP AAJ61888-AK CAAAJ61888-AP
BLOTTO Antifoam in PBS, with 0.02% Sodium Azide, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Blocking reagent for immunohistochemistry experiments.
  • Contains 5% nonfat dry milk, 0.01% Antifoam A , and 0.02% azide in PBS
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty