Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Angiotensin 1

Supplier: Thermo Scientific
J62102-MB J62102-LB0 J62102-LB0 J62102-MA J62102-MB
AAJ62102-MBEA 254.61 USD InStock
CAAAJ62102-MB CAAAJ62102-MA CAAAJ62102-LB0 AAJ62102-MB AAJ62102-LB0
Human Angiotensin 1
Proteins and Peptides
Powder

CAS No.: 484-42-4
Molecular Formula: C62H89N17O14
Formula Weight: 1296.50
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133091

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty