Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5(6)-Tetramethylrhodamine isothiocyanate

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: MRITCTetramethylrhodamine-5(6)-isothiocyanate5(6)-TRITCTRITC

J62258-MA J62258-MA
AAJ62258-MAEA 277.91 USD InStock
CAAAJ62258-MA AAJ62258-MA
5(6)-Tetramethylrhodamine isothiocyanate
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC]
Reagent for fluorescent labelling of proteins

Powder


Formula: C₂₅H₂₁N₃O₃S
MW: 443.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063517
CAS Number: 95197-95-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty