Would you like to log in again?
Clostridium histolyticum Collagenase, Type I
Supplier: Thermo Scientific
J62406-MC J62406-03 J62406-MC J62406-03
AAJ62406-MCEA 125.89 USD InStock
AAJ62406-MC AAJ62406-03 CAAAJ62406-MC CAAAJ62406-03
Clostridium histolyticum Collagenase, Type I
Enzymes
Applications: Isolated from Clostridium histolyticum. Degrades native helical collagen fibrils.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty