Would you like to log in again?
Streptavidin (from Streptomyces avidinii)
Supplier: Thermo Scientific
J62428-MA J62428-8PL J62428-MA J62428-8PL
AAJ62428-MAEA 556.14 USD InStock
AAJ62428-MA AAJ62428-8PL CAAAJ62428-8PL CAAAJ62428-MA
Streptavidin (from Streptomyces avidinii)
Proteins and Peptides
Streptavidin is used as a tetrameric protein to visualize biotin conjugated molecules in ELISA.
Due to the strong affinity of biotin and biotinylated molecules, streptavidin is one of the most important components in diagnostics and laboratory kits. Streptavidin is widely used in molecular biology through its unique high affinity for the biotin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty