Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TES sodium salt (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid sodium salt)

Supplier: Thermo Scientific Chemicals
Synonyms: TES sodium salt

J62706-30 J62706-14 J62706-14 J62706-A1
AAJ62706-14EA 66.91 USD InStock
CAAAJ62706-30 AAJ62706-14 CAAAJ62706-A1 CAAAJ62706-14
TES sodium salt (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid sodium salt)
TES sodium salt (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt)
A zwitterionic Good's Buffer

Powder


Formula: C₆H₁₄NNaO₆S
MW: 251.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065482
CAS Number: 70331-82-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty