Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calpain Inhibitor III

Supplier: Thermo Scientific
J62919-MB J62919-MB
AAJ62919-MBEA 152.82 USD InStock
AAJ62919-MB CAAAJ62919-MB
Calpain Inhibitor III
Proteins and Peptides
Powder

Purity: 95+%
CAS No.: 88191-84-8
Molecular Formula: C19H35N3O4S
Formula Weight: 401.60
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO at 25mg/ml
MDL No.: MFCD00798796

Applications: Inhibits calpain proteolysis

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty