Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trilaurin (glycerol trilaurate)

Supplier: Thermo Scientific Chemicals
Synonyms: Glycerol trilaurate Glyceryl trilaurate Glycerol tridodecanoate Glyceryl tridodecanoate

J62962-14 J62962-14 J62962-06
AAJ62962-14EA 82.06 USD InStock
CAAAJ62962-14 AAJ62962-14 AAJ62962-06
Trilaurin (glycerol trilaurate)
Trilaurin (Glycerol trilaurate)
Powder


Formula: C₃₉H₇₄O₆
MW: 639.01 g/mol
Melting Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty