Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-(2,3-Dihydroxypropyl)theophylline

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dyphylline

J63167-A1 J63167-A1 J63167-36 J63167-22 J63167-22 J63167-36
AAJ63167-36EA 339.7 USD InStock
CAAAJ63167-36 CAAAJ63167-A1 CAAAJ63167-22 AAJ63167-36 AAJ63167-A1 AAJ63167-22
7-(2,3-Dihydroxypropyl)theophylline
7-(2,3-Dihydroxypropyl)theophylline
An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator

Powder


Formula: C₁₀H₁₄N₄O₄
MW: 254.24 g/mol
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00005759
CAS Number: 479-18-5
EINECS: 207-526-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty