Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclosporin A

Supplier: Thermo Scientific
J63191-06 J63191-03 J63191-06 J63191-03
AAJ63191-06EA 494.02 USD InStock
AAJ63191-06 CAAAJ63191-03 CAAAJ63191-06 AAJ63191-03
Cyclosporin A
Proteins and Peptides
Crystalline solid

  • Soluble in DMSO or ethanol. Insoluble in water.

Signal word: Danger
GHS Symbols: GHS08, GHS07
Purity: 99+%
CAS No.: 59865-13-3
Molecular Formula: C62H111N11O12
Formula Weight: 1202.61
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Melting Point: 146-151°
Solubility: Soluble in DMSO or ethanol. Insoluble in water.
Beilstein Reference: 3647785
Merck Reference: 14,2752
MDL No.: MFCD00274558

Immunosuppressant. Induces apoptosis and inhibits angiogenesis induced by VEGF

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty