Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bradykinin (1-5)

Supplier: Thermo Scientific
J63233-LB0 J63233-MB J63233-LB0
AAJ63233-LB0EA 73.04 USD InStock
CAAAJ63233-MB CAAAJ63233-LB0 AAJ63233-LB0
Bradykinin (1-5)
Proteins and Peptides
Powder
Biologically active fragment of bradykinin

Molecular Formula: C27H40N8O6
Formula Weight: 572.67
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White

Biologically active fragment of bradykinin

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty