Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MG-132

Supplier: Thermo Scientific
J63250-LB0 J63250-MCR J63250-LB0 J63250-MA J63250-MA J63250-MCR
AAJ63250-MCREA 40 USD InStock
AAJ63250-MCR CAAAJ63250-LB0 CAAAJ63250-MCR AAJ63250-LB0 CAAAJ63250-MA AAJ63250-MA
MG-132
Proteins and Peptides
Crystalline solid

CAS No.: 133407-82-6
Molecular Formula: C26H41N3O5
Formula Weight: 475.63
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Appearance: White
MDL No.: MFCD00674886

Inhibitor of calpain proteases

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty