Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Native Bovine Trypsin (from Pancreas)

Supplier: Thermo Scientific
J63688-03 J63688-09 J63688-03 J63688-09
AAJ63688-03EA 52.94 USD InStock
AAJ63688-03 CAAAJ63688-03 AAJ63688-09 CAAAJ63688-09
Native Bovine Trypsin (from Pancreas)
Enzymes
A pancreatic serine protease.

  • CAS No.: 9002-07-7
  • EINECS: 232-650-8
  • GHS Symbol 1: GHS08
  • GHS Symbol 2: GHS07
  • Storage Temperature: -30°C to -10°C
  • Physical Form: Powder
  • Appearance: White to off-white
  • Merck Reference: 14,9795
  • MDL No.: MFCD00082094

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty