Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine Mucin (from Submaxillary Gland)

Supplier: Thermo Scientific
J63859-ME J63859-MC
AAJ63859-MEEA 461.94 USD InStock
AAJ63859-ME AAJ63859-MC
Bovine Mucin (from Submaxillary Gland)
Proteins and Peptides
Mucin is used in a study to investigate a galactose-specific lectin from the red marine alga Ptilota filicina.

  • Pale Brown Powder, 400 to 4000 kDa
  • Store at Cold Temperature
  • Form Suspension in water (pH 5.0)

Mucin from bovine submaxillary glands has also been used in a study to investigate the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line, neuraminidase substrate.

Caution: Incompatible with strong oxidizing agents.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty