Would you like to log in again?
BLOTTO Anti-foam in TBS, with 0.02% Sodium Azide, Thermo Scientific
Supplier: Thermo Scientific
J63891-AP J63891-AK J63891-AK J63891-AP
AAJ63891-APEA 97.6 USD InStock
AAJ63891-AP AAJ63891-AK CAAAJ63891-AK CAAAJ63891-AP
BLOTTO Anti-foam in TBS, with 0.02% Sodium Azide, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Blocking reagent for immunological procedure.
  • Contains 5% non-fat dry milk, 0.01% Antifoam A and 0.02% azide in TBS
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty