Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus)

Supplier: Thermo Scientific Chemicals
Synonyms: DNA sodium saltThymonucleic acid sodium salt

J64400-03 J64400-MC J64400-03 J64400-MC
AAJ64400-03EA 442.34 USD InStock
CAAAJ64400-MC CAAAJ64400-03 AAJ64400-03 AAJ64400-MC
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations.

  • White Lyophilized powder
  • Soluble in water
  • Store at cold temperatures

Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

Highly polymerized. Hyperchromicity is at least 27%.


Storage Temperature: Refrigerator
MDL Number: MFCD00130921
CAS Number: 9007-49-2
EINECS: 293-507-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty