Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sirtinol

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(((2-Hydroxynaphthalen-1-yl)methylene)amino)-N-(1-phenylethyl)benzamide

J64486-MA J64486-MA
AAJ64486-MAEA 212.04 USD InStock
AAJ64486-MA CAAAJ64486-MA
Sirtinol
Sirtinol
Sirtinol is a cell permeable specific inhibitor of sirtuin NAD-dependant deacetylases

Crystals


Formula: C₂₆H₂₂N₂O₂
MW: 394.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00810186
CAS Number: 410536-97-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty