Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

G3335

Supplier: Thermo Scientific
J64528-MC J64528-MF J64528-MC J64528-MF
AAJ64528-MFEA 127.83 USD InStock
CAAAJ64528-MC CAAAJ64528-MF AAJ64528-MF AAJ64528-MC
G3335
Proteins and Peptides
Solid

  • Light Sensitive

CAS No.: 36099-95-3
Molecular Formula: C16H19N3O5
Formula Weight: 333.34
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
MDL No.: MFCD00037964

Applications: A cell-permeable dipeptide that acts as a selective and reversible peroxisome proliferator-activated receptor antagonist

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty