Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromo-2'-deoxyguanosine 99%

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Warning
Synonyms: 2-Amino-8-bromo-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

J64723-03 J64723-03
AAJ64723-03EA 123.77 USD InStock
AAJ64723-03 CAAAJ64723-03
8-Bromo-2'-deoxyguanosine 99%
8-Bromo-2'-deoxyguanosine
Powder


Formula: C₁₀H₁₂BrN₅O₄
MW: 346.14 g/mol
Melting Pt: >250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01630971
CAS Number: 13389-03-2
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty